Assay Details
| Gene Symbol: Map2k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011943.2 | NP_036073.1 | 6 - 7 | 775 | 105 | |
| GenBank mRNA | AK043482.1 | - | 6 - 7 | 776 | 105 | |
| AK077712.1 | - | 6 - 7 | 770 | 105 | ||
| AK138757.1 | - | 6 - 7 | 776 | 105 | ||
| AK140289.1 | - | 7 - 8 | 760 | 105 | ||
| AK141708.1 | - | 6 - 7 | 760 | 105 | ||
| AK162671.1 | - | 6 - 7 | 729 | 105 | ||
| BC075652.1 | - | 6 - 7 | 774 | 30541969 | 105 | |
| U39066.1 | - | 6 - 7 | 627 | 105 | ||
| X97052.1 | - | 6 - 7 | 633 | 105 | ||
| X98067.1 | - | 6 - 7 | 868 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k6
Genomic Map