Assay Details
| Gene Symbol: Fbxo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153195.2 | NP_694875.2 | 2 - 3 | 511 | 81 | |
| GenBank mRNA | AK147797.1 | - | 2 - 3 | 463 | 81 | |
| AK147907.1 | - | 2 - 3 | 430 | 81 | ||
| AK150190.1 | - | 2 - 3 | 286 | 81 | ||
| AK150626.1 | - | 2 - 3 | 469 | 81 | ||
| AK151025.1 | - | 2 - 3 | 469 | 81 | ||
| AK152513.1 | - | 2 - 3 | 469 | 81 | ||
| AK182909.1 | - | 1 - 2 | 123 | 81 | ||
| BC032153.1 | - | 2 - 3 | 453 | 5054087 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo7
Genomic Map