Assay Details
| Gene Symbol: Fbxo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153195.2 | NP_694875.2 | 5 - 6 | 969 | 95 | |
| GenBank mRNA | AK010499.1 | - | 3 - 4 | 247 | 95 | |
| AK147716.1 | - | 269 | 95 | |||
| AK147797.1 | - | 5 - 6 | 921 | 95 | ||
| AK147907.1 | - | 5 - 6 | 888 | 95 | ||
| AK150190.1 | - | 5 - 6 | 744 | 95 | ||
| AK150626.1 | - | 5 - 6 | 927 | 95 | ||
| AK151025.1 | - | 5 - 6 | 927 | 95 | ||
| AK152513.1 | - | 5 - 6 | 927 | 95 | ||
| BC032153.1 | - | 5 - 6 | 911 | 5054087 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo7
Genomic Map