Assay Details
| Gene Symbol: Crispld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030209.3 | NP_084485.1 | 12 - 13 | 1427 | 59 | |
| GenBank mRNA | AK019034.1 | - | 12 - 13 | 1423 | 59 | |
| AK033858.1 | - | 12 - 13 | 1391 | 59 | ||
| AK132171.1 | - | 12 - 13 | 1370 | 59 | ||
| AK149909.1 | - | 12 - 13 | 1384 | 59 | ||
| AK149925.1 | - | 12 - 13 | 1384 | 59 | ||
| AK150180.1 | - | 12 - 13 | 1384 | 59 | ||
| AK154308.1 | - | 12 - 13 | 1427 | 59 | ||
| BC090642.1 | - | 12 - 13 | 1421 | 30653388 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crispld2
Genomic Map