Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
NM_001163760.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
NM_001163760.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
NM_001163761.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
NM_001163761.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
NM_001163762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
NM_001163762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
NM_172286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
NM_172286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011768.mm.1 |
TC0800001436.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011769.mm.1 |
NM_001163762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011769.mm.1 |
NM_001163762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011769.mm.1 |
TC0800001436.mm.1 |
MTA TC |