Assay Details
| Gene Symbol: 6430548M08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163761.1 | NP_001157233.1 | 9 - 10 | 1272 | 102 | |
| GenBank mRNA | AK133157.1 | - | 9 - 10 | 1270 | 102 | |
| AK172108.1 | - | 9 - 10 | 1273 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

6430548M08Rik
Genomic Map