Assay Details
| Gene Symbol: Taf1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021441.2 | NP_067416.2 | 6 - 7 | 595 | 116 | |
| GenBank mRNA | AK030522.1 | - | 6 - 7 | 562 | 116 | |
| AK087534.1 | - | 6 - 7 | 585 | 116 | ||
| AK154846.1 | - | 6 - 7 | 579 | 116 | ||
| AK156718.1 | - | 6 - 7 | 562 | 116 | ||
| BC058383.1 | - | 6 - 7 | 537 | 6811470 | 116 | |
| Y09974.1 | - | 6 - 7 | 722 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf1c
Genomic Map