Assay Details
| Gene Symbol: Adad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029428.1 | NP_083704.1 | 4 - 5 | 699 | 72 | |
| GenBank mRNA | AK015063.1 | - | 4 - 5 | 699 | 72 | |
| BC137980.1 | - | 4 - 5 | 686 | 8860999 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adad2
Genomic Map