| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800023259.mm.1 |
NM_001167910.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023259.mm.1 |
NM_001167910.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023259.mm.1 |
NM_019709.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023259.mm.1 |
NM_019709.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023259.mm.1 |
TC0800003022.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023258.mm.1 |
NM_001167910.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023258.mm.1 |
NM_001167910.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023258.mm.1 |
NM_019709.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023258.mm.1 |
NM_019709.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023258.mm.1 |
TC0800003022.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023255.mm.1 |
NM_001167910.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023255.mm.1 |
NM_001167910.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023255.mm.1 |
NM_019709.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023255.mm.1 |
NM_019709.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023255.mm.1 |
TC0800003022.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023256.mm.1 |
NM_001167910.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023256.mm.1 |
NM_001167910.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023256.mm.1 |
NM_019709.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023256.mm.1 |
NM_019709.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023256.mm.1 |
TC0800003022.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023254.mm.1 |
NM_001167910.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023254.mm.1 |
NM_001167910.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023254.mm.1 |
NM_019709.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023254.mm.1 |
NM_019709.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023254.mm.1 |
TC0800003022.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023257.mm.1 |
NM_019709.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023257.mm.1 |
NM_019709.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023257.mm.1 |
TC0800003022.mm.1 |
MTA TC |