Assay Details
| Gene Symbol: Tox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098799.1 | NP_001092269.1 | 4 - 5 | 685 | 64 | |
| GenBank mRNA | BF470404.1 | - | 2 - 3 | 481 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tox2
Genomic Map