Assay Details
| Gene Symbol: Abca8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013851.2 | NP_038879.2 | 5 - 6 | 771 | 113 | |
| GenBank mRNA | AF498362.1 | - | 5 - 6 | 771 | 113 | |
| AK138196.1 | - | 5 - 6 | 772 | 113 | ||
| AK173034.1 | - | 5 - 6 | 698 | 113 | ||
| BC089557.1 | - | 5 - 6 | 582 | 113 | ||
| BC141401.1 | - | 5 - 6 | 734 | 9056014 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca8b
Genomic Map