Assay Details
| Gene Symbol: Git1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004144.1 | NP_001004144.1 | 9 - 10 | 1012 | 68 | |
| GenBank mRNA | BC079870.1 | - | 9 - 10 | 1012 | 6831910 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Git1
Genomic Map