Assay Details
| Gene Symbol: Pipox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008952.2 | NP_032978.2 | 5 - 6 | 896 | 68 | |
| GenBank mRNA | AK008555.2 | - | 5 - 6 | 857 | 68 | |
| AK149488.1 | - | 5 - 6 | 845 | 68 | ||
| BC013525.1 | - | 5 - 6 | 810 | 4237443 | 68 | |
| U94700.1 | - | 5 - 6 | 808 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pipox
Genomic Map