Assay Details
| Gene Symbol: Engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172573.2 | NP_766161.1 | 11 - 12 | 1600 | 98 | |
| GenBank mRNA | AK048667.1 | - | 11 - 12 | 1602 | 98 | |
| AK084254.1 | - | 11 - 12 | 1733 | 98 | ||
| AK167142.1 | - | 11 - 12 | 1572 | 98 | ||
| AK171813.1 | - | 6 - 7 | 924 | 98 | ||
| BC120923.1 | - | 11 - 12 | 1598 | 98 | ||
| BC120924.1 | - | 11 - 12 | 1602 | 40097903 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Engase
Genomic Map