Assay Details
| Gene Symbol: Traf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009423.4 | NP_033449.2 | 3 - 4 | 409 | 58 | |
| GenBank mRNA | AF233449.1 | - | 3 - 4 | 308 | 58 | |
| AK035056.1 | - | 3 - 4 | 387 | 58 | ||
| AK040877.1 | - | 387 | 58 | |||
| AK046015.1 | - | 3 - 4 | 449 | 58 | ||
| AK053900.1 | - | 3 - 4 | 406 | 58 | ||
| BC020455.1 | - | 3 - 4 | 394 | 58 | ||
| X92346.1 | - | 3 - 4 | 429 | 58 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Traf4
Genomic Map