Assay Details
| Gene Symbol: Trp53i13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024920.1 | NP_001020091.1 | 5 - 6 | 559 | 76 | |
| GenBank mRNA | BC147324.1 | - | 5 - 6 | 557 | 9056369 | 76 |
| BC147325.1 | - | 5 - 6 | 557 | 9056377 | 76 | |
| BQ939386.1 | - | 5 - 6 | 559 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53i13
Genomic Map