Assay Details
| Gene Symbol: Trp53i13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024920.1 | NP_001020091.1 | 6 - 7 | 1089 | 59 | |
| GenBank mRNA | AK003215.1 | - | 1 - 2 | 359 | 59 | |
| BC147324.1 | - | 6 - 7 | 1087 | 9056369 | 59 | |
| BC147325.1 | - | 6 - 7 | 1087 | 9056377 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938072 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938073 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938074 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938059 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121372 | NM_001024920.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121372 | AK003215.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124649 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121372 | NM_001024920.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121372 | AK003215.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27938047 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07405670 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07405681 | - | - |
| Whole Mouse Genome G4122A | A_52_P35164 | NM_001024920.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P35164 | AK003215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P35164 | BC147324.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P35164 | BC147325.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07405670 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09159717 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09692965 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08987565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08987557 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09159715 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08987569 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08987568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08987572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938074 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938072 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938073 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27938055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27937974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27938042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09012360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09692967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09014543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09337012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09582717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09582711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27938054 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388676 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388674 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388672 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388675 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388673 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388677 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455315_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388669 | - | - |
| Mouse Genome 430 2.0 Array | 1455315_at | - | - |
| Mouse Genome 430 2.0 Array | 1455315_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216964_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216964_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100030100.mm.1 | NM_001024920.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030100.mm.1 | NM_001024920.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030100.mm.1 | TC1100003267.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100030098.mm.1 | NM_001024920.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030098.mm.1 | NM_001024920.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030098.mm.1 | TC1100003267.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100030099.mm.1 | NM_001024920.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030099.mm.1 | NM_001024920.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030099.mm.1 | TC1100003267.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455315_PM_at | - | - |
| Mouse Expression Set 430 | 1455315_at | - | - |
| Murine Genome U74v2 Set | 101432_PM_at | - | - |
| Murine Genome U74v2 Set | 101432_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455315_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455315_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455315_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455315_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 684572 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2758897 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2758897 | - | - |

Trp53i13
Genomic Map