Assay Details
| Gene Symbol: Rph3al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291159.1 | NP_001278088.1 | 4 - 5 | 519 | 79 | |
| GenBank mRNA | AK018335.1 | - | 4 - 5 | 519 | 79 | |
More Information
Set Membership: |
|

Rph3al
Genomic Map