Assay Details
| Gene Symbol: Nlrp1c-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027858.1 | 2 - 3 | 1707 | 117 | ||
| GenBank mRNA | BC141384.1 | - | 2 - 3 | 1703 | 9055997 | 117 |
| DQ117602.1 | - | 2 - 3 | 1707 | 117 | ||
More Information
Set Membership: |
|

Nlrp1c-ps
Genomic Map