Assay Details
| Gene Symbol: Kif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008444.4 | NP_032470.3 | 1 - 2 | 89 | 87 | |
| GenBank mRNA | AK081541.1 | - | 1 - 2 | 46 | 87 | |
| AK083700.1 | - | 1 - 2 | 77 | 87 | ||
| AK122258.1 | - | 1 - 2 | 50 | 87 | ||
| AK147467.1 | - | 1 - 2 | 83 | 87 | ||
| AK148200.1 | - | 1 - 2 | 70 | 87 | ||
| BC065132.1 | - | 1 - 2 | 56 | 87 | ||
| CJ089315.1 | - | 1 - 2 | 92 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083700.1, AK147467.1, AK148200.1, BC065132.1, NM_008444.4 transcripts.
Set Membership: |
|

Kif3b
Genomic Map