Assay Details
| Gene Symbol: Rnf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027258.1 | NP_081534.1 | 14 - 15 | 1775 | 80 | |
| GenBank mRNA | AK038042.1 | - | 8 - 9 | 1002 | 80 | |
| AK122571.1 | - | 4 - 5 | 511 | 80 | ||
| AK169566.1 | - | 9 - 10 | 955 | 80 | ||
| BC053070.1 | - | 14 - 15 | 1554 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf157
Genomic Map