| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100038945.mm.1 |
NM_027258.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038945.mm.1 |
NM_027258.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038945.mm.1 |
TC1100004115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100038946.mm.1 |
NM_027258.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038946.mm.1 |
NM_027258.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038946.mm.1 |
TC1100004115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100038949.mm.1 |
NM_027258.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038949.mm.1 |
NM_027258.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038949.mm.1 |
TC1100004115.mm.1 |
MTA TC |