Assay Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166506.1 | NP_001159978.1 | 9 - 10 | 1308 | 107 | |
| NM_001166507.1 | NP_001159979.1 | 9 - 10 | 1308 | 107 | ||
| NM_028777.3 | NP_083053.2 | 9 - 10 | 1308 | 107 | ||
| GenBank mRNA | AK004818.1 | - | 9 - 10 | 1233 | 107 | |
| AK047396.1 | - | 9 - 10 | 1316 | 107 | ||
| AK172947.1 | - | 6 - 7 | 843 | 107 | ||
| BC005766.1 | - | 9 - 10 | 1266 | 2649054 | 107 | |
| BC086620.1 | - | 9 - 10 | 1242 | 107 | ||
More Information
Set Membership: |
|

Sec14l1
Genomic Map