Assay Details
| Gene Symbol: Slc7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011404.3 | NP_035534.2 | 5 - 6 | 1034 | 72 | |
| GenBank mRNA | AB017189.1 | - | 5 - 6 | 980 | 72 | |
| AB023409.1 | - | 5 - 6 | 962 | 72 | ||
| AK142288.1 | - | 5 - 6 | 1035 | 72 | ||
| AK148134.1 | - | 5 - 6 | 704 | 72 | ||
| AK202552.1 | - | 3 - 4 | 193 | 72 | ||
| AK207202.1 | - | 3 - 4 | 224 | 72 | ||
| AK215643.1 | - | 3 - 4 | 223 | 72 | ||
| BC026131.1 | - | 5 - 6 | 1043 | 4188854 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a5
Genomic Map