Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800006341.mm.1 |
NM_001083906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006341.mm.1 |
NM_001083906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006341.mm.1 |
TC0800000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800006332.mm.1 |
NM_001083906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006332.mm.1 |
NM_001083906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006332.mm.1 |
TC0800000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800006335.mm.1 |
NM_001083906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006335.mm.1 |
NM_001083906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006335.mm.1 |
TC0800000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800006337.mm.1 |
NM_001083906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006337.mm.1 |
NM_001083906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006337.mm.1 |
TC0800000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800006336.mm.1 |
NM_001083906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006336.mm.1 |
NM_001083906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006336.mm.1 |
TC0800000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800006338.mm.1 |
NM_001083906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006338.mm.1 |
NM_001083906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800006338.mm.1 |
TC0800000854.mm.1 |
MTA TC |