Assay Details
| Gene Symbol: Nr3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083906.1 | NP_001077375.1 | 4 - 5 | 2049 | 60 | |
| GenBank mRNA | AK136580.1 | - | 1 - 2 | 92 | 60 | |
| BC133713.1 | - | 4 - 5 | 2049 | 40143429 | 60 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CV307991.1 | - | Chr. - - [-] on Build | 60 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nr3c2
Genomic Map