Assay Details
| Gene Symbol: Ddx42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028074.4 | NP_082350.3 | 13 - 14 | 1640 | 101 | |
| GenBank mRNA | AK049311.1 | - | 13 - 14 | 1642 | 101 | |
| AK134182.1 | - | 12 - 13 | 1446 | 101 | ||
| AK169816.1 | - | 13 - 14 | 1586 | 101 | ||
| AK171730.1 | - | 13 - 14 | 1509 | 101 | ||
| BC043036.1 | - | 12 - 13 | 1397 | 5698965 | 101 | |
| BC049382.1 | - | 1 - 2 | 52 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx42
Genomic Map