Assay Details
| Gene Symbol: Ddx42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028074.4 | NP_082350.3 | 5 - 6 | 713 | 125 | |
| GenBank mRNA | AK049311.1 | - | 5 - 6 | 715 | 125 | |
| AK169816.1 | - | 5 - 6 | 659 | 125 | ||
| AK171730.1 | - | 5 - 6 | 582 | 125 | ||
| BC043036.1 | - | 4 - 5 | 470 | 5698965 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx42
Genomic Map