Assay Details
| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054055.2 | NP_473396.1 | 11 - 12 | 1526 | 89 | |
| GenBank mRNA | AF306491.1 | - | 11 - 12 | 1525 | 89 | |
| AK085326.1 | - | 11 - 12 | 90 | |||
| AK128982.1 | - | 11 - 12 | 1523 | 89 | ||
| AK138344.1 | - | 11 - 12 | 1524 | 89 | ||
| AK165355.1 | - | 11 - 12 | 1529 | 89 | ||
| BC026803.1 | - | 11 - 12 | 1512 | 4238769 | 89 | |
| BC028882.1 | - | 10 - 11 | 1385 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc13a3
Genomic Map