Assay Details
| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054055.2 | NP_473396.1 | 8 - 9 | 1148 | 77 | |
| GenBank mRNA | AF306491.1 | - | 8 - 9 | 1147 | 77 | |
| AK085326.1 | - | 8 - 9 | 1146 | 77 | ||
| AK087518.1 | - | 8 - 9 | 1146 | 77 | ||
| AK128982.1 | - | 8 - 9 | 1145 | 77 | ||
| AK138344.1 | - | 8 - 9 | 1146 | 77 | ||
| AK157881.1 | - | 8 - 9 | 1117 | 77 | ||
| BC026803.1 | - | 8 - 9 | 1134 | 4238769 | 77 | |
| BC028882.1 | - | 7 - 8 | 1007 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc13a3
Genomic Map