Assay Details
| Gene Symbol: Snx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133924.3 | NP_598685.2 | 2 - 3 | 466 | 57 | |
| GenBank mRNA | AK141663.1 | - | 2 - 3 | 467 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819742 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819740 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819739 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | AK033724.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | AK161207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | BC017623.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | BC037155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | AK141663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | AK033724.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | AK161207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | BC017623.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | BC037155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323521 | AK141663.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168322 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177245 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122988 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148991 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | AK033724.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | AK161207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | BC017623.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | BC037155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | AK141663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | AK033724.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | AK161207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | BC017623.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | BC037155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323521 | AK141663.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819738 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122988 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04772998 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148991 | - | - |
| Whole Mouse Genome G4122A | A_52_P595561 | BC117542.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595561 | AK168473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595561 | AK017516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595561 | AK141663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595561 | NM_133924.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | AK033724.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | AK161207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | BC017623.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | BC037155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | AK141663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | AK033724.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | AK161207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | BC017623.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | BC037155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323521 | AK141663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P303523 | AK039166.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148991 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122988 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01420720 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819742 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819740 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01397647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01624707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01852842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01583139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01331123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01282904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02094821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01786488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819735 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10478586 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478583 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478584 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478585 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478581 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427070_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453776_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489553 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478580 | - | - |
| Mouse Genome 430 2.0 Array | 1453776_at | - | - |
| Mouse Genome 430 2.0 Array | 1453776_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427070_at | - | - |
| Mouse Genome 430 2.0 Array | 1427070_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101113_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101113_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200020713.mm.1 | NM_133924.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020713.mm.1 | NM_133924.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020713.mm.1 | TC0200002533.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200020712.mm.1 | NM_133924.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020712.mm.1 | NM_133924.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020712.mm.1 | TC0200002533.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200020711.mm.1 | NM_133924.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020711.mm.1 | NM_133924.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020711.mm.1 | TC0200002533.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200020714.mm.1 | NM_133924.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020714.mm.1 | NM_133924.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020714.mm.1 | TC0200002533.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200020703.mm.1 | NM_133924.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020703.mm.1 | NM_133924.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020703.mm.1 | TC0200002533.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427070_PM_at | - | - |
| Mouse Expression Set 430 | 1427070_at | - | - |
| Mouse Expression Set 430 | 1453776_PM_at | - | - |
| Mouse Expression Set 430 | 1453776_at | - | - |
| Murine Genome U74v2 Set | 104488_at | - | - |
| Murine Genome U74v2 Set | 104488_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427070_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427070_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427070_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427070_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453776_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453776_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 734098 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2491804 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2807449 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2807449 | - | - |

Snx21
Genomic Map