Assay Details
| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020333.2 | NP_065066.2 | 24 - 25 | 3540 | 69 | |
| GenBank mRNA | AF332063.1 | - | 24 - 25 | 3210 | 69 | |
| AF332064.1 | - | 24 - 25 | 3210 | 69 | ||
| AK122460.1 | - | 24 - 25 | 3274 | 69 | ||
| AK147262.1 | - | 24 - 25 | 3542 | 69 | ||
| BC054808.1 | - | 24 - 25 | 3206 | 6838880 | 69 | |
| KJ535321.1 | - | 24 - 25 | 3231 | 69 | ||
| KJ535322.1 | - | 24 - 25 | 3387 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a5
Genomic Map