Assay Details
| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146129.3 | NP_666241.1 | 7 - 8 | 979 | 72 | |
| GenBank mRNA | AK089358.1 | - | 7 - 8 | 980 | 72 | |
| AK150740.1 | - | 6 - 7 | 870 | 72 | ||
| BC031431.1 | - | 7 - 8 | 948 | 4241283 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcif1
Genomic Map