Assay Details
| Gene Symbol: Acot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133240.2 | NP_573503.2 | 3 - 4 | 548 | 67 | |
| GenBank mRNA | AF441166.1 | - | 3 - 4 | 548 | 67 | |
| AK033724.1 | - | 3 - 4 | 517 | 67 | ||
| AK151924.1 | - | 3 - 4 | 526 | 67 | ||
| BC005792.1 | - | 3 - 4 | 548 | 3603527 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot8
Genomic Map