Assay Details
| Gene Symbol: Acot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133240.2 | NP_573503.2 | 5 - 6 | 913 | 66 | |
| GenBank mRNA | AK151924.1 | - | 5 - 6 | 891 | 66 | |
| BC005792.1 | - | 5 - 6 | 913 | 3603527 | 66 | |
| BC052526.1 | - | 2 - 3 | 1047 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
acyl-CoA metabolic process
peroxisome organization
peroxisome fission
dicarboxylic acid catabolic process
receptor binding
palmitoyl-CoA hydrolase activity
hydrolase activity
choloyl-CoA hydrolase activity
acyl-CoA hydrolase activity
carboxylic ester hydrolase activity
medium-chain acyl-CoA hydrolase activity
acyl-CoA metabolic process
peroxisome organization
peroxisome fission
dicarboxylic acid catabolic process
receptor binding
palmitoyl-CoA hydrolase activity
hydrolase activity
choloyl-CoA hydrolase activity
acyl-CoA hydrolase activity
carboxylic ester hydrolase activity
medium-chain acyl-CoA hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819852 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038412 | BC005792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038412 | BC052526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038412 | NM_133240.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038412 | AF441166.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038412 | AI122325.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038414 | BC005792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038414 | NM_133240.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038414 | AF441166.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038414 | AI122325.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038414 | AK033724.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39480 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21727 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038412 | BC005792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038412 | BC052526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038412 | NM_133240.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038412 | AF441166.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038412 | AI122325.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038414 | BC005792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038414 | NM_133240.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038414 | AF441166.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038414 | AI122325.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038414 | AK033724.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P454078 | BC005792.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P454078 | BC052526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P454078 | NM_133240.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P454078 | AF441166.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P454078 | AI122325.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819829 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00550455 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00550450 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04773023 | - | - |
| Whole Mouse Genome G4122A | A_51_P454078 | BC005792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454078 | BC052526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454078 | NM_133240.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454078 | AF441166.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454078 | AI122325.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133939 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00550455 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01613402 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02070580 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02070582 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01333732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01988950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02024847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01613402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01160229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01160241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01160237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02070580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01160230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02024851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01883012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01160238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01464916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819834 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489559 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489560 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489557 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489558 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489561 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489555 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489556 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417449_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478580 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489553 | - | - |
| Mouse Genome 430 2.0 Array | 1417449_at | - | - |
| Mouse Genome 430 2.0 Array | 1417449_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170789_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170789_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200043745.mm.1 | NM_133240.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043745.mm.1 | NM_133240.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043745.mm.1 | TC0200005132.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417449_at | - | - |
| Mouse Expression Set 430 | 1417449_PM_at | - | - |
| Murine Genome U74v2 Set | 103386_PM_at | - | - |
| Murine Genome U74v2 Set | 103386_at | - | - |
| Murine Genome U74v2 Set | 166997_PM_at | - | - |
| Murine Genome U74v2 Set | 166997_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417449_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417449_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417449_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417449_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 823108 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2940039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2940039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5420025 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420025 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2738629 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250358 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2738629 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003112.1_0-S | - | - |
| mouse-6 v1.1 (Illumina) | scl18352.5.6_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420025 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18352.5.6_5-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003112.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940039 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420025 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2983587 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2738629 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1250358 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2983587 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2738629 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250358 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003112.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18352.5.6_5-S | - | - |

Acot8
Genomic Map