Assay Details
| Gene Symbol: Gipc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018771.3 | NP_061241.1 | 1 - 2 | 57 | 92 | |
| GenBank mRNA | AK086506.1 | - | 1 - 2 | 58 | 92 | |
| BC003490.1 | - | 1 - 2 | 57 | 2811749 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK086506.1, BC003490.1, NM_018771.3 transcripts.
Set Membership: |
|

Gipc1
Genomic Map