Assay Details
| Gene Symbol: Rap1gap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015046.2 | NP_001015046.1 | 18 - 19 | 1790 | 62 | |
| GenBank mRNA | AK038834.1 | - | 8 - 9 | 853 | 62 | |
| AK122424.1 | - | 4 - 5 | 583 | 62 | ||
| AK147290.1 | - | 19 - 20 | 1772 | 62 | ||
| BC107388.2 | - | 9 - 10 | 996 | 40057754 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rap1gap2
Genomic Map