Assay Details
| Gene Symbol: Dcaf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172502.3 | NP_766090.2 | 7 - 8 | 1234 | 101 | |
| GenBank mRNA | AK032982.1 | - | 7 - 8 | 1246 | 101 | |
| AK144808.1 | - | 7 - 8 | 1237 | 101 | ||
| AK153305.1 | - | 7 - 8 | 1225 | 101 | ||
| AK155000.1 | - | 7 - 8 | 1235 | 101 | ||
| BC057552.1 | - | 7 - 8 | 1235 | 6403172 | 101 | |
More Information
Set Membership: |
|

Dcaf15
Genomic Map