Assay Details
| Gene Symbol: Prmt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081240.3 | NP_001074709.1 | 7 - 8 | 1288 | 87 | |
| GenBank mRNA | AK045315.1 | - | 7 - 8 | 1278 | 87 | |
| AK154546.1 | - | 7 - 8 | 1232 | 87 | ||
| AK161023.1 | - | 7 - 8 | 1197 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prmt10
Genomic Map