Assay Details
| Gene Symbol: Arhgap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030113.2 | NP_084389.2 | 12 - 13 | 1290 | 91 | |
| GenBank mRNA | AF297029.1 | - | 12 - 13 | 1194 | 91 | |
| AF297030.1 | - | 12 - 13 | 1253 | 91 | ||
| AY179965.1 | - | 12 - 13 | 1166 | 91 | ||
| BU151977.1 | - | 2 - 3 | 116 | 91 | ||
| CN703944.1 | - | 5 - 6 | 477 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap10
Genomic Map