Assay Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159324.1 | NP_001152796.1 | 13 - 14 | 2061 | 58 | |
| NM_008064.3 | NP_032090.3 | 13 - 14 | 2151 | 58 | ||
| GenBank mRNA | AK052211.1 | - | 13 - 14 | 2132 | 58 | |
| AK088481.1 | - | 13 - 14 | 2127 | 58 | ||
| AK139333.1 | - | 13 - 14 | 2099 | 58 | ||
| AK141546.1 | - | 13 - 14 | 2134 | 58 | ||
| AK145631.1 | - | 12 - 13 | 1567 | 58 | ||
| AK146538.1 | - | 13 - 14 | 2055 | 58 | ||
| AK150970.1 | - | 13 - 14 | 2109 | 58 | ||
| AK191415.1 | - | 1 - 2 | 94 | 58 | ||
| AK204650.1 | - | 1 - 2 | 40 | 58 | ||
| BC010210.1 | - | 13 - 14 | 2115 | 3969127 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gaa
Genomic Map