Assay Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159324.1 | NP_001152796.1 | 9 - 10 | 1609 | 50 | |
| NM_008064.3 | NP_032090.3 | 9 - 10 | 1699 | 50 | ||
| GenBank mRNA | AK052211.1 | - | 9 - 10 | 1680 | 50 | |
| AK088481.1 | - | 9 - 10 | 1675 | 50 | ||
| AK139333.1 | - | 9 - 10 | 1647 | 50 | ||
| AK140042.1 | - | 9 - 10 | 1667 | 50 | ||
| AK141546.1 | - | 9 - 10 | 1682 | 50 | ||
| AK145631.1 | - | 8 - 9 | 1115 | 50 | ||
| AK146538.1 | - | 9 - 10 | 1603 | 50 | ||
| AK150970.1 | - | 9 - 10 | 1657 | 50 | ||
| AK212044.1 | - | 2 - 3 | 134 | 50 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gaa
Genomic Map