Assay Details
| Gene Symbol: Nptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008730.2 | NP_032756.1 | 2 - 3 | 877 | 90 | |
| GenBank mRNA | AK140128.1 | - | 2 - 3 | 394 | 90 | |
| BC093479.1 | - | 2 - 3 | 802 | 30947054 | 90 | |
| BC139018.1 | - | 2 - 3 | 784 | 8862040 | 90 | |
| BC139020.1 | - | 2 - 3 | 784 | 8862042 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nptx1
Genomic Map