| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700008216.mm.1 |
TC0700000908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008216.mm.1 |
NM_001037906.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008216.mm.1 |
NM_001037906.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008213.mm.1 |
TC0700000908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008213.mm.1 |
NM_001037906.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008213.mm.1 |
NM_001037906.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008214.mm.1 |
TC0700000908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008214.mm.1 |
NM_001037906.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008214.mm.1 |
NM_001037906.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008217.mm.1 |
TC0700000908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008217.mm.1 |
NM_001037906.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008217.mm.1 |
NM_001037906.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008215.mm.1 |
TC0700000908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008215.mm.1 |
NM_001037906.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008215.mm.1 |
NM_001037906.2 |
Refseq |