Assay Details
| Gene Symbol: Zfp335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199027.2 | NP_950192.2 | 10 - 11 | 1933 | 93 | |
| GenBank mRNA | AK157577.1 | - | 10 - 11 | 1764 | 93 | |
| BC062921.1 | - | 10 - 11 | 1933 | 93 | ||
| BC151013.1 | - | 9 - 10 | 1673 | 9087926 | 93 | |
| BC157954.1 | - | 9 - 10 | 1673 | 9088127 | 93 | |
| BC172135.1 | - | 9 - 10 | 1673 | 93 | ||
More Information
Set Membership: |
|

Zfp335
Genomic Map