Assay Details
| Gene Symbol: Zfp335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199027.2 | NP_950192.2 | 9 - 10 | 1823 | 94 | |
| GenBank mRNA | AK157577.1 | - | 9 - 10 | 1654 | 94 | |
| BC062921.1 | - | 9 - 10 | 1823 | 94 | ||
| BC151013.1 | - | 8 - 9 | 1563 | 9087926 | 94 | |
| BC157954.1 | - | 8 - 9 | 1563 | 9088127 | 94 | |
| BC172135.1 | - | 8 - 9 | 1563 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp335
Genomic Map