Assay Details
| Gene Symbol: Map1lc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026160.4 | NP_080436.1 | 1 - 2 | 93 | 75 | |
| GenBank mRNA | AF255953.1 | - | 1 - 2 | 111 | 75 | |
| AK002795.1 | - | 1 - 2 | 96 | 75 | ||
| AK003106.1 | - | 1 - 2 | 94 | 75 | ||
| AK003205.1 | - | 1 - 2 | 94 | 75 | ||
| AK003558.1 | - | 1 - 2 | 96 | 75 | ||
| AK012604.1 | - | 1 - 2 | 97 | 75 | ||
| AK132329.1 | - | 101 | 75 | |||
| AK151614.1 | - | 1 - 2 | 92 | 75 | ||
| BC068180.1 | - | 1 - 2 | 96 | 30634082 | 75 | |
| BC086774.1 | - | 1 - 2 | 80 | 75 | ||
| BC110308.1 | - | 1 - 2 | 84 | 5319360 | 75 | |
More Information
Set Membership: |
|

Map1lc3b
Genomic Map