Assay Details
| Gene Symbol: Map1lc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026160.4 | NP_080436.1 | 2 - 3 | 151 | 89 | |
| GenBank mRNA | AF255953.1 | - | 2 - 3 | 169 | 89 | |
| AK002795.1 | - | 2 - 3 | 154 | 89 | ||
| AK003106.1 | - | 2 - 3 | 152 | 89 | ||
| AK003205.1 | - | 2 - 3 | 152 | 89 | ||
| AK003558.1 | - | 2 - 3 | 154 | 89 | ||
| AK012604.1 | - | 2 - 3 | 155 | 89 | ||
| AK132329.1 | - | 159 | 89 | |||
| AK151614.1 | - | 2 - 3 | 150 | 89 | ||
| BC068180.1 | - | 2 - 3 | 154 | 30634082 | 89 | |
| BC110308.1 | - | 2 - 3 | 142 | 5319360 | 89 | |
More Information
Set Membership: |
|

Map1lc3b
Genomic Map