Assay Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138656.2 | NP_619597.2 | 5 - 6 | 662 | 57 | |
| NR_028354.1 | 6 - 7 | 765 | 57 | |||
| GenBank mRNA | AJ309922.1 | - | 5 - 6 | 619 | 57 | |
| AK089354.1 | - | 5 - 6 | 664 | 57 | ||
| AK134803.1 | - | 5 - 6 | 651 | 57 | ||
| AK153439.1 | - | 6 - 7 | 740 | 57 | ||
| AK154883.1 | - | 5 - 6 | 664 | 57 | ||
| AK170773.1 | - | 5 - 6 | 658 | 57 | ||
| BC008526.1 | - | 5 - 6 | 630 | 3495424 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvd
Genomic Map