Assay Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138656.2 | NP_619597.2 | 6 - 7 | 742 | 79 | |
| NR_028354.1 | 7 - 8 | 845 | 79 | |||
| GenBank mRNA | AK134803.1 | - | 6 - 7 | 731 | 79 | |
| AK153439.1 | - | 7 - 8 | 820 | 79 | ||
| AK170773.1 | - | 6 - 7 | 738 | 79 | ||
| BC008526.1 | - | 6 - 7 | 710 | 3495424 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvd
Genomic Map